Product Details

SNP ID
rs141498480
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:152971934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCAGCAGCAGCAGTGCCCAC[A/C]CCAGAGGGCCCAGCAGCAGCAAGTG
Phenotype
MIM: 616363
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 802 Missense Mutation CAC,CCC H15P NP_775103.1
XM_017000482.1 802 Missense Mutation CAC,CCC H15P XP_016855971.1

View Full Product Details