Product Details

SNP ID
rs141540345
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:90712579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGCCGAGTAGTTCCCTGCCTC[A/G]GCCAGCAACTCCAGGCCCACCGCTG
Phenotype
MIM: 605212
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BARHL2 PubMed Links

Gene Details

Gene
BARHL2
Gene Name
BarH like homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020063.1 939 Silent Mutation NP_064447.1

View Full Product Details