Product Details

SNP ID
rs141591555
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:39680820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGGGACCAAGCTCTATTGATCA[C/G]CATCCCCCAGAGTTTTCACCTGCAG
Phenotype
MIM: 610525
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HPCAL4 PubMed Links

Gene Details

Gene
HPCAL4
Gene Name
hippocalcin like 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282396.1 2705 UTR 3 NP_001269325.1
NM_001282397.1 2705 UTR 3 NP_001269326.1
NM_016257.3 2705 UTR 3 NP_057341.1
XM_017001447.1 2705 UTR 3 XP_016856936.1
Gene
NT5C1A
Gene Name
5'-nucleotidase, cytosolic IA
There are no transcripts associated with this gene.

View Full Product Details