Product Details

SNP ID
rs141851701
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:234373807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCAATCGCCTGCTTGGTGATT[A/G]TGGCCCCCACACACCTGTTTCTACA
Phenotype
MIM: 614772
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COA6 PubMed Links

Gene Details

Gene
COA6
Gene Name
cytochrome c oxidase assembly factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012985.2 125 Missense Mutation TAT,TGT Y30C NP_001013003.1
NM_001206641.2 125 Intron NP_001193570.2
NM_001301733.1 125 UTR 5 NP_001288662.1
Gene
LOC101927765
Gene Name
uncharacterized LOC101927765
There are no transcripts associated with this gene.

View Full Product Details