Product Details

SNP ID
rs141861416
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:9104470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTTGGGATTCCTGAGATTCTCC[C/T]GGCTTGGAAGGCGCGGGAGCCTTGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR157 PubMed Links

Gene Details

Gene
GPR157
Gene Name
G protein-coupled receptor 157
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024980.4 854 Silent Mutation CCA,CCG P319P NP_079256.4
XM_005263496.4 854 Silent Mutation CCA,CCG P306P XP_005263553.1
XM_005263497.4 854 Silent Mutation CCA,CCG P254P XP_005263554.1

View Full Product Details