Product Details

SNP ID
rs142052998
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:155210565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATCAGGAACTCTGCCTGCCCTT[C/T]GGACCAGTCATGGAGAGGTCATCTC
Phenotype
MIM: 600605 MIM: 188062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GBAP1 PubMed Links

Gene Details

Gene
GBAP1
Gene Name
glucosylceramidase beta pseudogene 1
There are no transcripts associated with this gene.

Gene
MTX1
Gene Name
metaxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002455.4 722 Missense Mutation CGG,TGG R206W NP_002446.3
NM_198883.3 722 Missense Mutation CGG,TGG R206W NP_942584.2
Gene
THBS3
Gene Name
thrombospondin 3
There are no transcripts associated with this gene.

View Full Product Details