Product Details

SNP ID
hCV166359280
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:206110207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTCCTAAAAGATGATGGTCTC[G/A]GCGGTGCCTTCCCCATCTGCCAGCT
Phenotype
MIM: 600264 MIM: 616088
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
AVPR1B PubMed Links

Gene Details

Gene
AVPR1B
Gene Name
arginine vasopressin receptor 1B
There are no transcripts associated with this gene.

Gene
C1orf186
Gene Name
chromosome 1 open reading frame 186
There are no transcripts associated with this gene.

View Full Product Details