Product Details
- SNP ID
-
rs142485597
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:108974752 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTACAGCTACAGATGCCACTGCAC[C/T]GCCTGTAGTGGTAGGAATGAAAGTT
- Phenotype
-
MIM: 615734
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR47
PubMed Links
Gene Details
- Gene
- WDR47
- Gene Name
- WD repeat domain 47
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142550.1 |
2775 |
Missense Mutation |
AGT,GGT |
S809G |
NP_001136022.1 |
NM_001142551.1 |
2775 |
Missense Mutation |
AGT,GGT |
S801G |
NP_001136023.1 |
NM_014969.5 |
2775 |
Missense Mutation |
AGT,GGT |
S802G |
NP_055784.3 |
XM_006710460.3 |
2775 |
Missense Mutation |
AGT,GGT |
S804G |
XP_006710523.1 |
XM_011541027.2 |
2775 |
Missense Mutation |
AGT,GGT |
S810G |
XP_011539329.1 |
XM_011541028.2 |
2775 |
Missense Mutation |
AGT,GGT |
S803G |
XP_011539330.1 |
XM_011541029.1 |
2775 |
Missense Mutation |
AGT,GGT |
S811G |
XP_011539331.1 |
XM_011541030.1 |
2775 |
Missense Mutation |
AGT,GGT |
S804G |
XP_011539332.1 |
XM_011541031.1 |
2775 |
Missense Mutation |
AGT,GGT |
S711G |
XP_011539333.1 |
XM_017000696.1 |
2775 |
Missense Mutation |
AGT,GGT |
S808G |
XP_016856185.1 |
XM_017000697.1 |
2775 |
Missense Mutation |
AGT,GGT |
S801G |
XP_016856186.1 |
View Full Product Details