Product Details

SNP ID
rs142760907
Assay Type
Functionally Tested
NCBI dbSNP Submissions
6
Location
Chr.1:74706958 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCGACTCCTTTGCCATGGTGTCTC[A/G]TGGGTTTATTTCAATAGTACCTCTG
Phenotype
MIM: 123691
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CRYZ PubMed Links
Additional Information
For this assay, SNP(s) [rs17552114] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CRYZ
Gene Name
crystallin zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130042.1 758 Nonsense Mutation CGA,TGA R257* NP_001123514.1
NM_001130043.1 758 Nonsense Mutation CGA,TGA R223* NP_001123515.1
NM_001134759.1 758 Nonsense Mutation CGA,TGA R120* NP_001128231.1
NM_001889.3 758 Nonsense Mutation CGA,TGA R257* NP_001880.2
XM_005270491.4 758 Nonsense Mutation CGA,TGA R120* XP_005270548.1
XM_011540747.1 758 Nonsense Mutation CGA,TGA R257* XP_011539049.1
XM_017000367.1 758 Nonsense Mutation CGA,TGA R223* XP_016855856.1
XM_017000368.1 758 Nonsense Mutation CGA,TGA R120* XP_016855857.1

View Full Product Details