Product Details
- SNP ID
-
rs142843575
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:160739321 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCAATATGGCTGGTTCCCCAACAT[A/G]CCTCACCCTCATCTATATCCTTTGG
- Phenotype
-
MIM: 606625
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLAMF7
PubMed Links
Gene Details
- Gene
- SLAMF7
- Gene Name
- SLAM family member 7
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282588.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269517.1 |
NM_001282589.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269518.1 |
NM_001282590.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269519.1 |
NM_001282591.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269520.1 |
NM_001282592.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269521.1 |
NM_001282593.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269522.1 |
NM_001282594.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269523.1 |
NM_001282595.1 |
265 |
Intron |
|
|
NP_001269524.1 |
NM_001282596.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_001269525.1 |
NM_021181.4 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
NP_067004.3 |
XM_011509828.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
XP_011508130.1 |
XM_011509829.1 |
265 |
Missense Mutation |
TAC,TGC |
Y7C |
XP_011508131.1 |
View Full Product Details