Product Details

SNP ID
rs142900519
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:54012161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCACGGCACAAGCTCTCATCCT[C/T]GTCCTCGCCGTGGGTACAGGTGCGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LDLRAD1 PubMed Links

Gene Details

Gene
LDLRAD1
Gene Name
low density lipoprotein receptor class A domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010978.3 267 Missense Mutation AAG,GAG K108E NP_001010978.2
NM_001276392.1 267 Missense Mutation AAG,GAG K69E NP_001263321.1
NM_001276393.1 267 Missense Mutation AAG,GAG K65E NP_001263322.1
NM_001276394.1 267 Intron NP_001263323.1
NM_001276395.1 267 Silent Mutation ACA,ACG T54T NP_001263324.1
XM_011541446.1 267 Missense Mutation AAG,GAG K107E XP_011539748.1
XM_017001264.1 267 Silent Mutation ACA,ACG T54T XP_016856753.1

View Full Product Details