Product Details

SNP ID
rs142910641
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:247605622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGAGAGTTCCCTAATGGATTTC[A/G]TCCTTCTAGGCTTCTCAGACCACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2G3 PubMed Links

Gene Details

Gene
OR2G3
Gene Name
olfactory receptor family 2 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001914.1 37 Missense Mutation ATC,GTC I13V NP_001001914.1

View Full Product Details