Product Details

SNP ID
rs143264882
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:235120216 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGCCATTATTGTAGATTTTTATA[C/T]AACTGTAAGCAATTTGTCCTTATGA
Phenotype
MIM: 601848
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNORA14B PubMed Links

Gene Details

Gene
SNORA14B
Gene Name
small nucleolar RNA, H/ACA box 14B
There are no transcripts associated with this gene.

Gene
TOMM20
Gene Name
translocase of outer mitochondrial membrane 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014765.2 Intron NP_055580.1

View Full Product Details