Product Details

SNP ID
rs143334205
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:155054178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGCTGCTACCAGGGAAGAGTGC[A/G]GGGATATGCAGGCTCCTGGGTGTCC
Phenotype
MIM: 605548 MIM: 601380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM15 PubMed Links

Gene Details

Gene
ADAM15
Gene Name
ADAM metallopeptidase domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261464.1 486 Missense Mutation CAG,CGG Q134R NP_001248393.1
NM_001261465.1 486 Missense Mutation CAG,CGG Q124R NP_001248394.1
NM_001261466.1 486 Missense Mutation CAG,CGG Q108R NP_001248395.1
NM_003815.4 486 Missense Mutation CAG,CGG Q124R NP_003806.3
NM_207191.2 486 Missense Mutation CAG,CGG Q124R NP_997074.1
NM_207194.2 486 Missense Mutation CAG,CGG Q124R NP_997077.1
NM_207195.2 486 Missense Mutation CAG,CGG Q124R NP_997078.1
NM_207196.2 486 Missense Mutation CAG,CGG Q124R NP_997079.1
NM_207197.2 486 Missense Mutation CAG,CGG Q124R NP_997080.1
Gene
DCST1
Gene Name
DC-STAMP domain containing 1
There are no transcripts associated with this gene.

Gene
EFNA4
Gene Name
ephrin A4
There are no transcripts associated with this gene.

Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details