Product Details

SNP ID
rs143447407
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:111449863 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCTCTGAAGAATGCAGCCTTC[C/T]TAGGTCCAGGGTAAGTGTGAGGATA
Phenotype
MIM: 603270 MIM: 611734
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP5F1 PubMed Links

Gene Details

Gene
ATP5F1
Gene Name
ATP synthase, H+ transporting, mitochondrial Fo complex subunit B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001688.4 488 Silent Mutation CTA,TTA L23L NP_001679.2
Gene
WDR77
Gene Name
WD repeat domain 77
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317062.1 488 Intron NP_001303991.1
NM_001317063.1 488 Intron NP_001303992.1
NM_001317064.1 488 Intron NP_001303993.1
NM_024102.3 488 Intron NP_077007.1

View Full Product Details