Product Details

SNP ID
rs143716190
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:117957146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAACTCTTTAACGAATTCATTCA[A/G]CTGGGCTCTGATGTTGAACTTATAT
Phenotype
MIM: 616554 MIM: 604737
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPAG17 PubMed Links

Gene Details

Gene
SPAG17
Gene Name
sperm associated antigen 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206996.2 2598 Intron NP_996879.1
XM_006710426.3 2598 Intron XP_006710489.1
XM_006710427.3 2598 Intron XP_006710490.1
XM_006710428.3 2598 Intron XP_006710491.1
XM_006710429.3 2598 Intron XP_006710492.1
XM_006710431.3 2598 Intron XP_006710494.1
XM_011540934.1 2598 Intron XP_011539236.1
XM_011540935.2 2598 Intron XP_011539237.1
XM_011540936.2 2598 Intron XP_011539238.1
XM_011540937.2 2598 Intron XP_011539239.1
XM_011540939.2 2598 Intron XP_011539241.1
XM_011540941.2 2598 Intron XP_011539243.1
XM_011540942.2 2598 Intron XP_011539244.1
Gene
WDR3
Gene Name
WD repeat domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006784.2 2598 Silent Mutation CAA,CAG Q844Q NP_006775.1

View Full Product Details