Product Details

SNP ID
rs143746483
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:205715047 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCACGATTCCCATCCCCAACCA[C/G]TACTTGAAAATCCACTTATCTGAAT
Phenotype
MIM: 611912
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NUCKS1 PubMed Links

Gene Details

Gene
NUCKS1
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022731.4 4209 UTR 3 NP_073568.2
XM_005245453.1 4209 UTR 3 XP_005245510.1

View Full Product Details