Product Details
- SNP ID
-
rs144473968
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
8
- Location
-
Chr.1:45345250 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCAGTCAGCCAGGGGCATAGTTC[C/T]GGGCCCTGGTGCATCCAGGTCAAAT
- Phenotype
-
MIM: 604933
MIM: 604746
MIM: 613931
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MUTYH
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3893383] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MUTYH
- Gene Name
- mutY DNA glycosylase
There are no transcripts associated with this gene.
- Gene
- TESK2
- Gene Name
- testis-specific kinase 2
- Gene
- TOE1
- Gene Name
- target of EGR1, member 1 (nuclear)
There are no transcripts associated with this gene.
View Full Product Details