Product Details

SNP ID
rs144665333
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:999705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCCGACTTACCTCTTTTCTGAGG[A/G]CGTCCAGGATGAGGGTTTTGAGCTG
Phenotype
MIM: 608060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HES4 PubMed Links

Gene Details

Gene
HES4
Gene Name
hes family bHLH transcription factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142467.1 468 Missense Mutation GCC,GTC A90V NP_001135939.1
NM_021170.3 468 Missense Mutation GCC,GTC A64V NP_066993.1
XM_005244771.4 468 Intron XP_005244828.1

View Full Product Details