Product Details

SNP ID
rs144988372
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:155053418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACCTGCCCTCTGGTGCCCACAGA[C/G]CAGTCTGCCTGAGCCCCTGAGGATC
Phenotype
MIM: 605548
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAM15 PubMed Links

Gene Details

Gene
ADAM15
Gene Name
ADAM metallopeptidase domain 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261464.1 303 Missense Mutation ACC,AGC T73S NP_001248393.1
NM_001261465.1 303 Missense Mutation ACC,AGC T63S NP_001248394.1
NM_001261466.1 303 Intron NP_001248395.1
NM_003815.4 303 Missense Mutation ACC,AGC T63S NP_003806.3
NM_207191.2 303 Missense Mutation ACC,AGC T63S NP_997074.1
NM_207194.2 303 Missense Mutation ACC,AGC T63S NP_997077.1
NM_207195.2 303 Missense Mutation ACC,AGC T63S NP_997078.1
NM_207196.2 303 Missense Mutation ACC,AGC T63S NP_997079.1
NM_207197.2 303 Missense Mutation ACC,AGC T63S NP_997080.1
Gene
DCST1
Gene Name
DC-STAMP domain containing 1
There are no transcripts associated with this gene.

Gene
LOC100505666
Gene Name
uncharacterized LOC100505666
There are no transcripts associated with this gene.

View Full Product Details