Product Details

SNP ID
rs145190932
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:156009609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACCACAATAGCAGGGGGATGCC[A/G]ATGGAGGGAAGGGTCATGACCCCAA
Phenotype
MIM: 600867
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SSR2 PubMed Links

Gene Details

Gene
SSR2
Gene Name
signal sequence receptor subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003145.3 563 Silent Mutation ATC,ATT I161I NP_003136.1
XM_011509923.2 563 Silent Mutation ATC,ATT I180I XP_011508225.1

View Full Product Details