Product Details

SNP ID
rs145229745
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:247606068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTAGTTCCCTAATCCATGCAAC[C/T]TTTACCTTGCAATTGCCTCTCTGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR2G3 PubMed Links

Gene Details

Gene
OR2G3
Gene Name
olfactory receptor family 2 subfamily G member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001914.1 483 Silent Mutation ACC,ACT T161T NP_001001914.1

View Full Product Details