Product Details

SNP ID
rs145315934
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:153608082 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTCTTGACCAGGCTGTACTTAG[A/G]CACATATTTGTAGAAGTTTTCCACC
Phenotype
MIM: 607986
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
S100A14 PubMed Links

Gene Details

Gene
S100A14
Gene Name
S100 calcium binding protein A14
There are no transcripts associated with this gene.

Gene
S100A16
Gene Name
S100 calcium binding protein A16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317007.1 313 Missense Mutation CCT,TCT P24S NP_001303936.1
NM_001317008.1 313 Missense Mutation CCT,TCT P24S NP_001303937.1
NM_080388.2 313 Missense Mutation CCT,TCT P24S NP_525127.1

View Full Product Details