Product Details

SNP ID
rs145533453
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:109115645 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAACTCCATCCCATATGGAGATAA[A/G]TTCTTATAAAGATGTTCTGTGTCTA
Phenotype
MIM: 611298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf194 PubMed Links

Gene Details

Gene
C1orf194
Gene Name
chromosome 1 open reading frame 194
There are no transcripts associated with this gene.

Gene
KIAA1324
Gene Name
KIAA1324
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267048.1 Intron NP_001253977.1
NM_001284352.1 Intron NP_001271281.1
NM_001284353.1 Intron NP_001271282.1
NM_020775.4 Intron NP_065826.2
XM_011541825.1 Intron XP_011540127.1
XM_011541826.2 Intron XP_011540128.1
XM_011541827.1 Intron XP_011540129.1
XM_017001883.1 Intron XP_016857372.1

View Full Product Details