Product Details

SNP ID
rs145645598
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:229326870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACTCGATGATCTTTGGGGACT[G/T]TTGACCGCTTTCCTGTTTCCTCTAG
Phenotype
MIM: 616762
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCSAP PubMed Links

Gene Details

Gene
CCSAP
Gene Name
centriole, cilia and spindle associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145257.4 582 Missense Mutation AAA,AAC K168N NP_660300.3

View Full Product Details