Product Details

SNP ID
rs145787587
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:42849697 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCAACCCACTCTGCTGAAATGTC[A/G]TTATTTCTTCAAGGCCCGGAGGAAA
Phenotype
MIM: 609017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
There are no transcripts associated with this gene.

Gene
LOC339539
Gene Name
uncharacterized LOC339539
There are no transcripts associated with this gene.

Gene
ZNF691
Gene Name
zinc finger protein 691
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242739.1 3022 Silent Mutation TCA,TCG S13S NP_001229668.1
NM_015911.3 3022 Intron NP_056995.1
XM_006710671.3 3022 Intron XP_006710734.1
XM_017001400.1 3022 Silent Mutation TCA,TCG S13S XP_016856889.1
XM_017001401.1 3022 Silent Mutation TCA,TCG S13S XP_016856890.1
XM_017001402.1 3022 UTR 5 XP_016856891.1
XM_017001403.1 3022 UTR 5 XP_016856892.1

View Full Product Details