Product Details

SNP ID
rs145791121
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:231529319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACATAACCAAAGAAGAGAAGGGAA[A/G]GATGTTAATTCATCTTCACCCGTGA
Phenotype
MIM: 602964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSNAX PubMed Links

Gene Details

Gene
TSNAX
Gene Name
translin associated factor X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005999.2 239 Silent Mutation AAA,AAG K27K NP_005990.1
Gene
TSNAX-DISC1
Gene Name
TSNAX-DISC1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details