Product Details

SNP ID
rs145863585
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:153816465 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGTGAGTTGGCAGCATCAGTCA[C/T]GGCGCTGGGGCTAGGAAGTGGCGAG
Phenotype
MIM: 614998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GATAD2B PubMed Links

Gene Details

Gene
GATAD2B
Gene Name
GATA zinc finger domain containing 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005245364.4 1119 Missense Mutation ATG,GTG M342V XP_005245421.1

View Full Product Details