Product Details

SNP ID
rs146301626
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:31183979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTCTCCATCCACCAGGAGCGGTG[C/T]AGGGATGTGTTGAAGGTCATGATGA
Phenotype
MIM: 612871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NKAIN1 PubMed Links

Gene Details

Gene
NKAIN1
Gene Name
Na+/K+ transporting ATPase interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024522.2 649 Silent Mutation CTA,CTG L103L NP_078798.2
XM_017002320.1 649 Silent Mutation CTA,CTG L76L XP_016857809.1

View Full Product Details