Product Details

SNP ID
rs146456111
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:150733109 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGATTTCCTTTATTTCTTGCCAT[A/C]CGAATATATCCTTCTTCACCAAAGT
Phenotype
MIM: 116845
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTSS PubMed Links

Gene Details

Gene
CTSS
Gene Name
cathepsin S
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199739.1 1194 Silent Mutation CGG,CGT R261R NP_001186668.1
NM_004079.4 1194 Silent Mutation CGG,CGT R311R NP_004070.3

View Full Product Details