Product Details

SNP ID
rs146461576
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:224190202 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTATAGCTGAGCATTACATGTT[C/G]TTAAAGGGTCATGAAACTTACTCAT
Phenotype
MIM: 615843
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DEGS1 PubMed Links

Gene Details

Gene
DEGS1
Gene Name
delta 4-desaturase, sphingolipid 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321541.1 837 Missense Mutation TTC,TTG F236L NP_001308470.1
NM_001321542.1 837 Missense Mutation TTC,TTG F200L NP_001308471.1
NM_003676.3 837 Missense Mutation TTC,TTG F236L NP_003667.1
XM_017002648.1 837 Missense Mutation TTC,TTG F200L XP_016858137.1

View Full Product Details