Product Details

SNP ID
rs146547241
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:31310103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGCCATAGAATATTAAGGATGA[A/C]TTCAGGAAGGCCTGAGACCATGGAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZCCHC17 PubMed Links

Gene Details

Gene
ZCCHC17
Gene Name
zinc finger CCHC-type containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282566.1 228 Intron NP_001269495.1
NM_001282567.1 228 Intron NP_001269496.1
NM_001282568.1 228 Missense Mutation AAT,ACT N2T NP_001269497.1
NM_001282569.1 228 UTR 5 NP_001269498.1
NM_001282570.1 228 Intron NP_001269499.1
NM_001282571.1 228 UTR 5 NP_001269500.1
NM_001282572.1 228 Missense Mutation AAT,ACT N2T NP_001269501.1
NM_001282573.1 228 UTR 5 NP_001269502.1
NM_001282574.1 228 Missense Mutation AAT,ACT N2T NP_001269503.1
NM_016505.3 228 Missense Mutation AAT,ACT N2T NP_057589.2
XM_006710681.3 228 UTR 5 XP_006710744.1
XM_011541567.2 228 Missense Mutation AAT,ACT N2T XP_011539869.1

View Full Product Details