Product Details

SNP ID
rs146574652
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:32363619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTTTATCCAGAGATTCCTCCCTC[A/G]GGAGCTCCAAATCGTCCGTACCCTG
Phenotype
MIM: 607660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BSDC1 PubMed Links

Gene Details

Gene
BSDC1
Gene Name
BSD domain containing 1
There are no transcripts associated with this gene.

Gene
FAM229A
Gene Name
family with sequence similarity 229 member A
There are no transcripts associated with this gene.

Gene
TSSK3
Gene Name
testis specific serine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052841.3 611 Missense Mutation CAG,CGG Q57R NP_443073.1

View Full Product Details