Product Details

SNP ID
rs146621405
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:46310213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTGGAGAAATGTGTAAAGGCCC[A/G]GGAGCGGCTAGAGCTCTGTGATGAG
Phenotype
MIM: 613844
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRRC41 PubMed Links

Gene Details

Gene
LRRC41
Gene Name
leucine rich repeat containing 41
There are no transcripts associated with this gene.

Gene
UQCRH
Gene Name
ubiquinol-cytochrome c reductase hinge protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297565.1 294 Missense Mutation CAG,CGG Q41R NP_001284494.1
NM_001297566.1 294 Missense Mutation CAG,CGG Q38R NP_001284495.1
NM_006004.3 294 Missense Mutation CAG,CGG Q47R NP_005995.2

View Full Product Details