Product Details

SNP ID
rs146627789
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:77946775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGCAAAATACTGCAATTGTTCA[C/T]GTCAATACCCTACTCTTAAAGATAA
Phenotype
MIM: 603444 MIM: 613121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FUBP1 PubMed Links

Gene Details

Gene
FUBP1
Gene Name
far upstream element binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303433.1 4092 Intron NP_001290362.1
NM_003902.4 4092 Intron NP_003893.2
XM_011542391.2 4092 Intron XP_011540693.1
XM_011542392.2 4092 Intron XP_011540694.1
XM_011542393.2 4092 Intron XP_011540695.1
XM_017002739.1 4092 Intron XP_016858228.1
XM_017002740.1 4092 UTR 3 XP_016858229.1
XM_017002741.1 4092 Intron XP_016858230.1
XM_017002742.1 4092 UTR 3 XP_016858231.1
XM_017002743.1 4092 UTR 3 XP_016858232.1
XM_017002744.1 4092 UTR 3 XP_016858233.1
Gene
NEXN
Gene Name
nexilin F-actin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172309.1 4092 Intron NP_001165780.1
NM_144573.3 4092 Intron NP_653174.3
XM_005271322.3 4092 Intron XP_005271379.1
XM_005271323.3 4092 Intron XP_005271380.1
XM_005271324.4 4092 Intron XP_005271381.1
XM_005271325.3 4092 Intron XP_005271382.1
XM_005271326.3 4092 Intron XP_005271383.1
XM_005271327.3 4092 Intron XP_005271384.1

View Full Product Details