Product Details

SNP ID
rs146726959
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:151365214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGACCACTAGGGGCTCTGGCTGG[C/G]ACTTTAGTTCCTCGTCCTCCAGCAC
Phenotype
MIM: 604188
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SELENBP1 PubMed Links

Gene Details

Gene
SELENBP1
Gene Name
selenium binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258288.1 1414 Missense Mutation TCC,TGC S309C NP_001245217.1
NM_001258289.1 1414 Missense Mutation TCC,TGC S413C NP_001245218.1
NM_003944.3 1414 Missense Mutation TCC,TGC S371C NP_003935.2
XM_017002757.1 1414 Intron XP_016858246.1

View Full Product Details