Product Details

SNP ID
rs147360157
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:152972058 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGTCAAGAAGCAATGCCCACC[A/G]AAAGGCACCATCATTCCAGCCCAGC
Phenotype
MIM: 616363
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPRR4 PubMed Links

Gene Details

Gene
SPRR4
Gene Name
small proline rich protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173080.2 926 Silent Mutation CCA,CCG P56P NP_775103.1
XM_017000482.1 926 Silent Mutation CCA,CCG P56P XP_016855971.1

View Full Product Details