Product Details

SNP ID
rs147599277
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:31183838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTGCCAGGAAGATCTGCAGGGC[A/G]CTGCTGAGGGCTTCAATGTAGGGGT
Phenotype
MIM: 612871
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKAIN1 PubMed Links

Gene Details

Gene
NKAIN1
Gene Name
Na+/K+ transporting ATPase interacting 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024522.2 790 Silent Mutation AGC,AGT S150S NP_078798.2
XM_017002320.1 790 Silent Mutation AGC,AGT S123S XP_016857809.1

View Full Product Details