Product Details

SNP ID
rs147708219
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:40251507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATCGTTGCTCAAAAACCTGCCAC[A/G]AAGAGGGATTGCTCCTCTGAGCCCT
Phenotype
MIM: 606480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMCO2 PubMed Links

Gene Details

Gene
TMCO2
Gene Name
transmembrane and coiled-coil domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008740.3 555 Silent Mutation ACA,ACG T154T NP_001008740.1
Gene
ZMPSTE24
Gene Name
zinc metallopeptidase STE24
There are no transcripts associated with this gene.

View Full Product Details