Product Details

SNP ID
rs147721587
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:205718334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACCCTTCATCCCCAGATTTCTC[A/G]GGTGGGGGGCTTGTAGATGTTTTCT
Phenotype
MIM: 611912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUCKS1 PubMed Links

Gene Details

Gene
NUCKS1
Gene Name
nuclear casein kinase and cyclin dependent kinase substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022731.4 922 Silent Mutation CCC,CCT P226P NP_073568.2
XM_005245453.1 922 Silent Mutation CCC,CCT P225P XP_005245510.1

View Full Product Details