Product Details
- SNP ID
-
rs147728519
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
11
- Location
-
Chr.1:32633978 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGTTTCTACTTCTACTGTTTGGA[A/G]GGGCTCCACTGTCCCAGTATCTGTC
- Phenotype
-
MIM: 615891
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZBTB8OS
PubMed Links
Gene Details
- Gene
- ZBTB8OS
- Gene Name
- zinc finger and BTB domain containing 8 opposite strand
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001308135.1 |
257 |
Missense Mutation |
CTC,TTC |
L78F |
NP_001295064.1 |
NM_001308136.1 |
257 |
Missense Mutation |
CTC,TTC |
L85F |
NP_001295065.1 |
NM_001308137.1 |
257 |
Missense Mutation |
CTC,TTC |
L78F |
NP_001295066.1 |
NM_001308138.1 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
NP_001295067.1 |
NM_001308139.1 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
NP_001295068.1 |
NM_001308140.1 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
NP_001295069.1 |
NM_001308141.1 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
NP_001295070.1 |
NM_178547.3 |
257 |
Missense Mutation |
CTC,TTC |
L85F |
NP_848642.1 |
XM_011541327.2 |
257 |
Missense Mutation |
CTC,TTC |
L85F |
XP_011539629.1 |
XM_011541328.1 |
257 |
Intron |
|
|
XP_011539630.1 |
XM_011541329.2 |
257 |
Missense Mutation |
CTC,TTC |
L37F |
XP_011539631.1 |
XM_011541330.2 |
257 |
Missense Mutation |
CTC,TTC |
L33F |
XP_011539632.1 |
XM_011541334.2 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
XP_011539636.1 |
XM_011541335.2 |
257 |
Intron |
|
|
XP_011539637.1 |
XM_011541337.2 |
257 |
Intron |
|
|
XP_011539639.1 |
XM_011541338.2 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
XP_011539640.1 |
XM_017001134.1 |
257 |
Intron |
|
|
XP_016856623.1 |
XM_017001135.1 |
257 |
Missense Mutation |
CTC,TTC |
L85F |
XP_016856624.1 |
XM_017001136.1 |
257 |
Intron |
|
|
XP_016856625.1 |
XM_017001137.1 |
257 |
Intron |
|
|
XP_016856626.1 |
XM_017001138.1 |
257 |
Missense Mutation |
CTC,TTC |
L16F |
XP_016856627.1 |
View Full Product Details