Product Details

SNP ID
rs147916945
Assay Type
Functionally Tested
NCBI dbSNP Submissions
3
Location
Chr.1:32077175 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCCCGACCCAGGCCTCTCAGAG[A/G]GGGAAGGTCTCCCTCTTTCGCTCCA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR5585 PubMed Links

Gene Details

Gene
MIR5585
Gene Name
microRNA 5585
There are no transcripts associated with this gene.

Gene
TMEM39B
Gene Name
transmembrane protein 39B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319677.1 865 Silent Mutation AGA,AGG R22R NP_001306606.1
NM_001319678.1 865 Intron NP_001306607.1
NM_001319679.1 865 Intron NP_001306608.1
NM_018056.3 865 Silent Mutation AGA,AGG R149R NP_060526.2
XM_005270988.2 865 Silent Mutation AGA,AGG R22R XP_005271045.1
XM_006710724.1 865 Silent Mutation AGA,AGG R149R XP_006710787.1
XM_006710725.3 865 Silent Mutation AGA,AGG R149R XP_006710788.1
XM_011541681.2 865 Silent Mutation AGA,AGG R149R XP_011539983.1
XM_011541683.1 865 Silent Mutation AGA,AGG R22R XP_011539985.1
XM_011541684.2 865 Intron XP_011539986.1
XM_017001595.1 865 Silent Mutation AGA,AGG R34R XP_016857084.1

View Full Product Details