Product Details

SNP ID
rs148005568
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:183563285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTCTGAAAGGGGCACCAGCTC[A/G]TTGCTGTCCCGAGGCCGATAGCTGG
Phenotype
MIM: 608515 MIM: 610964
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCF2 PubMed Links

Gene Details

Gene
NCF2
Gene Name
neutrophil cytosolic factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000433.3 1475 Silent Mutation AAC,AAT N400N NP_000424.2
NM_001127651.2 1475 Silent Mutation AAC,AAT N400N NP_001121123.1
NM_001190789.1 1475 Silent Mutation AAC,AAT N319N NP_001177718.1
NM_001190794.1 1475 Silent Mutation AAC,AAT N355N NP_001177723.1
XM_005245207.1 1475 Silent Mutation AAC,AAT N364N XP_005245264.1
XM_011509580.1 1475 Silent Mutation AAC,AAT N400N XP_011507882.1
XM_011509581.1 1475 Silent Mutation AAC,AAT N400N XP_011507883.1
Gene
SMG7
Gene Name
SMG7, nonsense mediated mRNA decay factor
There are no transcripts associated with this gene.

View Full Product Details