Product Details

SNP ID
rs148240494
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:2187823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGCCCAGGGCACCGTCAGTCCC[A/G]TGGGTGTGGACAGGCACGTGACAGT
Phenotype
MIM: 615183 MIM: 176982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAAP20 PubMed Links

Gene Details

Gene
FAAP20
Gene Name
Fanconi anemia core complex associated protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146310.1 Intron NP_001139782.1
NM_001256945.1 Intron NP_001243874.1
NM_001256946.1 Intron NP_001243875.1
NM_001256947.1 Intron NP_001243876.1
NM_001282670.1 Intron NP_001269599.1
NM_001282671.1 Intron NP_001269600.1
NM_001282672.1 Intron NP_001269601.1
NM_001282673.1 Intron NP_001269602.1
NM_182533.2 Intron NP_872339.2
XM_006710419.3 Intron XP_006710482.1
XM_006710421.3 Intron XP_006710484.1
XM_011540914.2 Intron XP_011539216.1
XM_011540921.2 Intron XP_011539223.1
XM_011540922.1 Intron XP_011539224.1
XM_017000553.1 Intron XP_016856042.1
XM_017000554.1 Intron XP_016856043.1
XM_017000555.1 Intron XP_016856044.1
XM_017000556.1 Intron XP_016856045.1
XM_017000557.1 Intron XP_016856046.1
Gene
LOC100506504
Gene Name
uncharacterized LOC100506504
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
There are no transcripts associated with this gene.

View Full Product Details