Product Details
- SNP ID
-
rs148244341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:9037933 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGATCAAGCCCACGGTGAAGTT[A/G]GAGAGCCAGTGCACACTGCCCCCCA
- Phenotype
-
MIM: 138230
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SLC2A5
PubMed Links
Gene Details
- Gene
- SLC2A5
- Gene Name
- solute carrier family 2 member 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135585.1 |
1487 |
Intron |
|
|
NP_001129057.1 |
NM_003039.2 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
NP_003030.1 |
XM_005263491.3 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_005263548.1 |
XM_017002133.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857622.1 |
XM_017002134.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857623.1 |
XM_017002135.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857624.1 |
XM_017002136.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857625.1 |
XM_017002137.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857626.1 |
XM_017002138.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857627.1 |
XM_017002139.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857628.1 |
XM_017002140.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857629.1 |
XM_017002141.1 |
1487 |
Silent Mutation |
TCC,TCT |
S422S |
XP_016857630.1 |
XM_017002142.1 |
1487 |
Silent Mutation |
TCC,TCT |
S363S |
XP_016857631.1 |
View Full Product Details