Product Details

SNP ID
rs148442336
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:28593762 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTCTTTGATGTGACAAACAGGA[A/G]GTCCTTTGAACACATCCAAGACTGG
Phenotype
MIM: 600773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB42 PubMed Links

Gene Details

Gene
RAB42
Gene Name
RAB42, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193532.1 319 Missense Mutation NP_001180461.1
NM_152304.1 319 Intron NP_689517.1
XM_017000226.1 319 Intron XP_016855715.1
Gene
TAF12
Gene Name
TATA-box binding protein associated factor 12
There are no transcripts associated with this gene.

View Full Product Details