Product Details

SNP ID
rs148596588
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:34851056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTAGATCCCCACCTCACCAGG[A/G]TCACAAGGATACACACAACTATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMIM12 PubMed Links

Gene Details

Gene
SMIM12
Gene Name
small integral membrane protein 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164824.1 5092 UTR 3 NP_001158296.1
NM_001164825.1 5092 UTR 3 NP_001158297.1
NM_001320261.1 5092 UTR 3 NP_001307190.1
NM_138428.5 5092 UTR 3 NP_612437.3
XM_005270403.4 5092 Intron XP_005270460.1
XM_011540553.2 5092 Intron XP_011538855.1
XM_011540554.2 5092 Intron XP_011538856.2
XM_011540555.2 5092 Intron XP_011538857.1
XM_017000166.1 5092 Intron XP_016855655.1

View Full Product Details