Product Details

SNP ID
rs148707904
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:151763039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAGGTTTTCTTTGGGCCGATG[C/G]TTCGTGTCCTCCACCAGCTTATAGA
Phenotype
MIM: 611824 MIM: 605138 MIM: 609501
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL9 PubMed Links

Gene Details

Gene
MRPL9
Gene Name
mitochondrial ribosomal protein L9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300733.1 698 Missense Mutation AAC,AAG N87K NP_001287662.1
NM_031420.3 698 Missense Mutation AAC,AAG N87K NP_113608.1
Gene
OAZ3
Gene Name
ornithine decarboxylase antizyme 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134939.1 698 UTR 5 NP_001128411.1
NM_001301371.1 698 Intron NP_001288300.1
NM_016178.2 698 Intron NP_057262.2
Gene
TDRKH
Gene Name
tudor and KH domain containing
There are no transcripts associated with this gene.

View Full Product Details