Product Details

SNP ID
rs148842291
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:23193574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCCCGGTAGATCCGGCCATATA[A/G]GATGATGAGCAACACCGAGGGAATG
Phenotype
MIM: 182133
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HTR1D PubMed Links

Gene Details

Gene
HTR1D
Gene Name
5-hydroxytryptamine receptor 1D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000864.4 1156 Silent Mutation CTA,TTA L216L NP_000855.1

View Full Product Details