Product Details
- SNP ID
-
rs148863520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:248521787 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTGGGGAACACTGCCCTCATACT[A/G]GTATGTTGTCTGGACTCCAGACTCC
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
OR2G6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs58955396] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OR2G6
- Gene Name
- olfactory receptor family 2 subfamily G member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013355.1 |
141 |
Silent Mutation |
CTA,CTG |
L47L |
NP_001013373.1 |
- Gene
- OR2T10
- Gene Name
- olfactory receptor family 2 subfamily T member 10
View Full Product Details